H-Arg(Me)-OH · HCl
Supplier: BACHEM AMERICAS INC
4028190.0050
E-2770.0050BAEA
304.81
USD
E-2770.0050BA
H-Arg(Me)-OH · HCl
H-Arg(Me)-OH · HCl
Sequence: H-Arg(Me)-OH · HCl
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).
Synonym(s): L-NMMA · HCl#L-NMA · HCl#Tilarginine · HCl#Targinine · HCl
Formula:
C₇H₁₆N₄O₂·HCl Storage Temperature: Freezer |
MDL Number:
MFCD01632059 CAS Number: 156706-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...