AC-ASP-NH₂

4010792.0050
E-2700.0050BAEA 131.52 USD
E-2700.0050BA
AC-ASP-NH₂
AC-ASP-NH₂

Sequence: Ac-Asp-NH₂


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Synonym(s): Ac-Isoasn-OH#Ac-L-isoasparagine

Formula: C₆H₁₀N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00237475
CAS Number: 60803-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR