L-Serine ethyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Ser-OEt·HCl

4013411.0100 4013411.0025
E-2680.0100BAEA 453.57 USD InStock
E-2680.0100BA E-2680.0025BA
L-Serine ethyl ester hydrochloride
L-Serine ethyl ester hydrochloride

Sequence: H-Ser-OEt · HCl


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₅H₁₂ClNO₃
MW: 169,605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012594
CAS Number: 26348-61-8
EINECS: 247-624-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR