BACEE-2605

4011720.0005 4011720.0001
E-2605.0001BAEA 148.58 USD
E-2605.0001BA E-2605.0005BA
BACEE-2605
Sequence: Ac-Tyr(Me)-OMe

Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
p-Methoxyphenylalanine and p-ethoxyphenylalanine.

Synonym(s): Ac-4-methoxy-Phe-OMe
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR