L-Cystine bisamide dihydrochloride

Synonyms: (H-Cys-NH₂)₂·2 HCl

4011703.0250 4011703.1000
E-2380.0250BAEA 175.34 USD
E-2380.0250BA E-2380.1000BA
L-Cystine bisamide dihydrochloride
L-Cystine bisamide dihydrochloride

Sequence: (H-Cys-NH₂)₂ · 2 HCl


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.

Formula: C₆H₁₆Cl₂N₄O₂S₂
MW: 311.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237450
CAS Number: 22671-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR