2-Amino-N,N-dimethylacetamide acetate

Supplier: BACHEM AMERICAS INC

Synonyms: H-Gly-NMe₂·acetate

4001603.0001
E-1985.0001BAEA 167.67 USD
E-1985.0001BA
2-Amino-N,N-dimethylacetamide acetate
2-Amino-N,N-dimethylacetamide acetate

Sequence: H-Gly-NMe₂ · acetate


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₆H₁₄N₂O₃
MW: 162.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237954
CAS Number: 200634-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR