Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycine benzyl ester-p-toluenesulfonate

Supplier: BACHEM AMERICAS INC

Synonyms: H-Gly-OBzl.Tos-OH

E-1975.0025 E-1975.0100
E-1975.0025BAEA 83.64 USD InStock
E-1975.0025BA E-1975.0100BA
Glycine benzyl ester-p-toluenesulfonate
Glycine benzyl ester-p-toluenesulfonate
Sequence: H-Gly-OBzl · p-tosylate

Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.


Formula: C₉H₁₁NO₂
MW: 337,39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00035425
CAS Number: 1738-76-7
EINECS: 217-094-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty