H-Glu(ANILIDE)-OH
Supplier: BACHEM AMERICAS INC
4005313.0005
4005313.0001
E-1935.0001BAEA
137.17
USD
E-1935.0001BA
E-1935.0005BA
H-Glu(ANILIDE)-OH
H-Glu(ANILIDE)-OH
Sequence: H-Glu(anilide)-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
See also the glutamic acid γ-diazomethyl ketones L-DON (F-1510) and D-DON (F-2185).
Synonym(s): H-Gln(Ph)-OH
Formula:
C₁₁H₁₄N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00063105 CAS Number: 5963-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...