Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-leucine

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Leu-OH

E-1100.0100 E-1100.0025
E-1100.0100BAEA 298.4 USD InStock
E-1100.0100BA E-1100.0025BA
N-Acetyl-L-leucine
N-Acetyl-L-leucine
Sequence: Ac-Leu-OH

Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).


Formula: C₈H₁₅NO₃
MW: 173,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065131
CAS Number: 1188-21-2
EINECS: 214-706-3
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty