H-Ser(tBu)-2-chlorotrityl resin 200-400 mesh

Supplier: BACHEM AMERICAS INC

4037969.0001 4037969.0005
D-2665.0001BAEA 450.73 USD
D-2665.0001BA D-2665.0005BA
H-Ser(tBu)-2-chlorotrityl resin 200-400 mesh
H-Ser(tBu)-2-chlorotrityl resin

Sequence: H-Ser(tBu)-2-chlorotrityl resin (200-400 mesh)
CAS-No.:
Mol.weight:
SumFormula:
Bulk item no.: D-2665
Synonym(s):
EINECS no.:
MDL no.: MFCD00801416
Substitution: 0.50-0.90 mmol/g

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR