Iodopolystyrene resin 200-400 mesh
4027178.0001
4027178.0005
D-2260.0001BAEA
148.58
USD
D-2260.0001BA
D-2260.0005BA
Iodopolystyrene resin 200-400 mesh
Iodopolystyrene resin
Sequence: Iodopolystyrene resin (200-400 mesh)
Synonym(s): I-Ph-polymer
This "polymeric iodide" has been oxidized to yield iodosyl compounds such as the iodosyl acetate (M.L.Hallensleben). Being an excellent leaving group, iodine may also be replaced with a metal (e.g. lithium). A broad range of resin derivatives can be obtained via metallated polymer (see e.g. M.J.Farrall and J.M.J.Fréchet).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...