BACED-2225

4027171.0001 4027171.0005
D-2225.0001BAEA 173.35 USD
D-2225.0001BA D-2225.0005BA
BACED-2225
Sequence: Isocyanomethyl resin (200-400 mesh)
Synonym(s): CN-CH₂-Ph-polymer

This "polymeric isonitrile" can be employed in multicomponent condensations such as the Passerini or the Ugi reaction.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR