Fmoc-D-Arg(Pbf)-Wang resin 0.50-1.00 mmol/g 200-400 mesh
4025776.0001
D-1995.0001BAEA
344.47
USD
D-1995.0001BA
Fmoc-D-Arg(Pbf)-Wang resin 0.50-1.00 mmol/g 200-400 mesh
Fmoc-D-Arg(Pbf)-Wang resin
Sequence: Fmoc-D-Arg(Pbf)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-D-Arg(Pbf)-p-alkoxybenzyl alcohol resin
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...