N-Fmoc-L-norleucine

Synonyms: Fmoc-Nle-OHFMOC-L-Norleucine

4018558.0001
D-1795.0001BAEA 96.52 USD
D-1795.0001BA
N-Fmoc-L-norleucine
N-Fmoc-L-norleucine

Sequence: Fmoc-Nle-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Nle-p-alkoxybenzyl alcohol resin#Fmoc-L-2-aminohexanoic acid-Wang resin#Fmoc-L-2-aminohexanoic acid-p-alkoxybenzyl alcohol resin

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037537
CAS Number: 77284-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR