Fmoc-Gln(Trt) 0.4 - 0.7 mmol/g on Wang resin 200-400 mesh
4016163.0001
4016163.0005
D-1690.0001BAEA
165.33
USD
D-1690.0001BA
D-1690.0005BA
Fmoc-Gln(Trt) 0.4 - 0.7 mmol/g on Wang resin 200-400 mesh
Fmoc-Gln(Trt)
Sequence: Fmoc-Gln(Trt)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Gln(Trt)-p-alkoxybenzyl alcohol resin
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...