Fmoc-Gln(Trt) 0.3-0.6 mmol/g on SASRIN™ resin 200-400 mesh
4016126.0001
D-1685.0001BAEA 358.69 USD
D-1685.0001BA
Fmoc-Gln(Trt) 0.3-0.6 mmol/g on SASRIN™ resin 200-400 mesh
Fmoc-Gln(Trt)
Sequence: Fmoc-Gln(Trt)-SASRIN™ resin (200-400 mesh)