BACED-1560

4014398.0001 4014398.0005
D-1560.0005BAEA 953.42 USD
D-1560.0005BA D-1560.0001BA
BACED-1560
Sequence: Fmoc-D-Arg(Pmc)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-D-Arg(Pmc)-p-alkoxybenzyl alcohol resin
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR