BACED-1485

4013477.0005 4013477.0025
D-1485.0025BAEA 569.57 USD
D-1485.0025BA D-1485.0005BA
BACED-1485
Sequence: 4-Hydroxymethyl-benzoyloxymethyl resin (200-400 mesh)
Synonym(s): 4-HOCH₂-Ph-COO-CH₂-Ph-polymer

C-terminally modified peptides can be obtained from this carrier by cleavage with nucleophiles.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR