Boc-Arg(Tos)-Merrifield resin 200-400 mesh

Supplier: BACHEM AMERICAS INC

4013551.0005 4013551.0025
D-1465.0005BAEA 131.31 USD
D-1465.0005BA D-1465.0025BA
Boc-Arg(Tos)-Merrifield resin 200-400 mesh
Boc-Arg(Tos)-Merrifield resin

Sequence: Boc-Arg(Tos)-Merrifield resin (200-400 mesh)
CAS-No.:
Mol.weight:
SumFormula:
Bulk item no.: D-1465
Synonym(s):
EINECS no.:
MDL no.: MFCD00801160
Substitution: 0.3-0.6 mmol/g

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR