Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloromethyl polystyrene resin 200-400 mesh

Supplier: BACHEM AMERICAS INC
Synonyms: Chloromethylated styrene-divinylbenzene copolymerMerrifield Resin

D-1245.0100 D-1245.0025
Merrifield resin is one of the standard supports for the synthesis of peptides by Boc strategy. Originally, the cesium salt of a protected amino acid was anchored to the chloromethyl support via nucleophilic displacement of chlorine. Me₄N salts, sodium salts in THF with Bu₄NF catalysis, and zinc salts in EtOH have been used as well. Cleavage is normally effected by treatment with HF or TFMSA or by catalytic transfer hydrogenolysis, alcohols can be released by using diisobutylaluminium hydride or LiBH₄. For cleavage scavengers may be required. New isomeric trialkoxybenzylamine resins have been developed by coupling phthalimidomethyl-3,5-dimethoxyphenols to this resin followed by subsequent treatment with hydrazine, that allows DCC coupling with the carboxyl function of amino acids or peptides. The addition of alcohols can be performed by heating the corresponding potassium or sodium alkoxide and the resin in DMF. Reductive cleavage of the anchoring ester linkage was also accomplished. Attachment of sodium dihydrocinnamate to this resin and treatment with a solution of lithium diisopropylamide (LDA) in THF formed a resin-bound zinc enolate for aldol condensations. Reductive cleavage with diisobutylaluminium hydride afforded a 1,3-diol library. The removal of β-lactams has been achieved by using aluminium chloride. Another application of this support to nonpeptides involved anchoring the cesium salts of bromo or iodobenzoic acids followed by subsequent transformation into biaryl derivatives using Suzuki coupling conditions. The products were released from the resin via nucleophilic attack with methoxide ion to give methyl esters. Bachem offers a large variety of Boc-amino acids linked to Merrifield resin.

  • Substitution: 0.80 - 1.40 mmol/g


Storage Temperature: Ambient
MDL Number: MFCD00146406
CAS Number: 55844-94-5


 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 g D-1245.0100 D-1245.0100BA Each (100g) Retrieving Restricted
25 g D-1245.0025 D-1245.0025BA Each (25g) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty