BACEC-3870

4025913.0005 4025913.0001
C-3870.0001BAEA 173.3 USD
C-3870.0001BA C-3870.0005BA
BACEC-3870
Sequence: Z-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Tic represents a constrained Phe analog.

Synonym(s): Z-D-Tic-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR