BACEC-3790

4025235.0005 4025235.0001
C-3790.0001BAEA 173.35 USD
C-3790.0001BA C-3790.0005BA
BACEC-3790
Sequence: Z-Aad(OtBu)-OH · DCHA
Synonym(s): Z-L-α-aminoadipic acid-δ-t-butyl ester · DCHA#(S)-2-(Z-amino)-hexanedioic acid-6-t-butyl ester · DCHA#Z-Homoglu(OtBu)-OH · DCHA
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR