BACEC-3785

4025232.0005 4025232.0001
C-3785.0005BAEA 397.47 USD
C-3785.0005BA C-3785.0001BA
BACEC-3785
Sequence: Z-Aad-OH
Synonym(s): Z-L-α-aminoadipic acid#(S)-2-(Z-amino)-hexanedioic acid#Z-Homoglu-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR