BACEC-3725

4008695.0001 4008695.0005
C-3725.0001BAEA 86.77 USD
C-3725.0001BA C-3725.0005BA
BACEC-3725
Sequence: Z-Nle-OSu
Synonym(s): Z-L-2-aminohexanoic acid-N-hydroxysuccinimide ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR