Z-2-Nal-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Z-L-2-Nal-OH

4015633.0005
C-3500.0005BAEA 1754.19 USD
C-3500.0005BA
Z-2-Nal-OH
Z-2-Nal-OH

Sequence: Z-2-Nal-OH


Fluorescent phenylalanine analog.

Formula: C₂₁H₁₉NO₄
MW: 349.386 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063338
CAS Number: 65365-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR