(S)-1-Benzyl-2-(2,5-dioxopyrrolidin-1-yl)-5-oxopyrrolidine-1,2-dicarboxylate

Synonyms: (S)-1-Benzyl 2-(2,5-dioxopyrrolidin-1-yl) 5-oxopyrrolidine-1,2-dicarboxylateN-Benzyloxycarbonyl-L-pyroglutamic acid N-succinimidyl esterZ-Pyr-OSu

4015205.0025 4015205.0005
C-3335.0025BAEA 344.47 USD
C-3335.0025BA C-3335.0005BA
(S)-1-Benzyl-2-(2,5-dioxopyrrolidin-1-yl)-5-oxopyrrolidine-1,2-dicarboxylate
(S)-1-Benzyl-2-(2,5-dioxopyrrolidin-1-yl)-5-oxopyrrolidine-1,2-dicarboxylate

Sequence: Z-Pyr-OSu


Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.

Formula: C₁₇H₁₆N₂O₇
MW: 360.322 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00191144
CAS Number: 40291-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR