Z-Tyr(tBu)-OMe

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-Methyl 2-(((benzyloxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoateN-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester

4000137.0005
C-3065.0005BAEA 124.95 USD
C-3065.0005BA
Z-Tyr(tBu)-OMe
Z-Tyr(tBu)-OMe

Sequence: Z-Tyr(tBu)-OMe


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₂H₂₇NO₅
MW: 385.46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237370
CAS Number: 5068-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR