BACEC-1950
4002867.0001 4002867.0005
C-1950.0001BAEA 84.2 USD
C-1950.0001BA C-1950.0005BA
BACEC-1950
Sequence: Z-D-His-OH
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.