Z-L-Asp(tBu)-ONp

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Asp(OtBu)-ONp

4002881.0100 4002881.0005 4002881.0025
C-1365.0100BAEA 1600.61 USD
C-1365.0100BA C-1365.0005BA C-1365.0025BA
Z-L-Asp(tBu)-ONp
Z-L-Asp(tBu)-ONp

Sequence: Z-Asp(OtBu)-ONp


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₂H₂₄N₂O₈
MW: 444,44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00235866
CAS Number: 17543-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR