Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Cbz-L-arginine hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Arg-OH·HClN-α-Cbz-L-Arginine Hydrochloride

C-1290.0100 C-1290.0025
C-1290.0100BAEA 312.72 USD InStock
C-1290.0100BA C-1290.0025BA
Cbz-L-arginine hydrochloride
Cbz-L-arginine hydrochloride
Sequence: Z-Arg-OH · HCl

Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.


Formula: C₁₄H₂₁ClN₄O₄
MW: 344,796 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038946
CAS Number: 56672-63-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty