Z-Ala-Lys-OH

Supplier: BACHEM AMERICAS INC

4002755.0005 4002755.0001
C-1140.0005BAEA 716.16 USD
C-1140.0005BA C-1140.0001BA
Z-Ala-Lys-OH
Z-Ala-Lys-OH

Sequence: Z-Ala-Lys-OH
CAS-No.: 76264-07-8
Mol.weight: 351.40
SumFormula: C₁₇H₂₅N₃O₅
Bulk item no.: C-1140
Synonym(s):
EINECS no.: -
MDL no.: MFCD00238404

Formula: C₁₇H₂₅N₃O₅
MW: 351.403 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238404
CAS Number: 76264-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR