Fmoc-4-(7-hydroxy-4-coumarinyl)-Abu-OH

Supplier: BACHEM AMERICAS INC

4076431.1000
B-4205.1000BAEA 2685.57 USD
B-4205.1000BA
Fmoc-4-(7-hydroxy-4-coumarinyl)-Abu-OH
Fmoc-4-(7-hydroxy-4-coumarinyl)-Abu-OH

Sequence: Fmoc-4-(7-hydroxy-4-coumarinyl)-Abu-OH
Synonym(s): Fmoc-(7-hydroxycoumarin-4-yl)-ethyl-Gly-OH#Fmoc-(umbellifer-4-yl)-ethyl-Gly-OH


Protected derivative of the fluorescent amino acid Cgl. Homolog of the coumarinyl alanine derivative B-3740.

Formula: C₂₈H₂₃NO₇
Storage Temperature: Freezer
MDL Number: MFCD22377745
CAS Number: 1187744-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR