Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Fmoc-(Dmb)Gly-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-(Dmb)Gly-OH

B-4170.0250 B-4170.1000 B-4170.5000
B-4170.0250BAEA 75.19 USD InStock
B-4170.0250BA B-4170.1000BA B-4170.5000BA
Fmoc-(Dmb)Gly-OH
Fmoc-(Dmb)Gly-OH
Derivative for the Fmoc-SPPS of backbone-protected peptides. Introduction of Dmb at appropriate positions prevents aggregation and aspartimide formation.Please see also our Dmb-dipeptides.

  • Sequence: Fmoc-(Dmb)Gly-OH
  • CAS-No.: 166881-42-1
  • Mol.weight: 447.49
  • SumFormula: C₂₆H₂₅NO₆
  • Bulk item no.: B-4170
  • Synonym(s): Fmoc-N-(2,4-dimethoxybenzyl)-Gly-OH
  • EINECS no.: -
  • MDL no.: MFCD08064316

Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.


Formula: C₂₆H₂₅NO₆
MW: 447,487 g/mol
CAS Number: 166881-42-1
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty