Fmoc-D-propargylglycine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Pra-OH

4072399.0001 4072399.0005
B-4150.0001BAEA 1434.9 USD
B-4150.0001BA B-4150.0005BA
Fmoc-D-propargylglycine
Fmoc-D-propargylglycine

Building block for the synthesis of precursor peptides for tritiation, and for use in "click chemistry".D-Pra-containing peptides can be modified or cyclized by 1,3-dipolar cycloaddition, e.g. with alkyl azides. Fmoc-D-Pra-OH can be used for obtaining aryl-modified D-Phe derivatives via rhodium-catalyzed [2+2+2]-cycloaddition.


  • Sequence: Fmoc-D-Pra-OH
  • Synonym(s): Fmoc-Propargyl-D-Gly-OH

Formula: C₂₀H₁₇NO₄
MW: 335.359 g/mol
MDL Number: MFCD01311781
CAS Number: 220497-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR