Fmoc-δ-azido-Nva-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Orn(N3)

4072395.0250 4072395.1000
B-4140.0250BAEA 227.9 USD
B-4140.0250BA B-4140.1000BA
Fmoc-δ-azido-Nva-OH
Fmoc-δ-azido-Nva-OH

Ornithine-derived building block for "click chemistry" and other reactions involving azides.The azido moiety may also be selectively converted into an amino group with triphenylphosphine in the presence of water (Staudinger reaction).


  • Sequence: Fmoc-δ-azido-Nva-OH
  • Synonym(s): Fmoc-Orn(N₂)-OH

Formula: C₂₀H₂₀N₄O₄
MW: 380.40 g/mol
CAS Number: 1097192-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR