Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Fmoc-L-Lys(N3)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-ε-azido-Nle-OH

B-4135.0250 B-4135.1000
B-4135.0250BAEA 347.33 USD InStock
B-4135.0250BA B-4135.1000BA
Fmoc-L-Lys(N3)-OH
Fmoc-L-Lys(N3)-OH
Lysine-derived building block for "click chemistry" and other reactions involving azides.The azido moiety of Lys(N₃) may also be selectively converted into an amino group with triphenylphosphine in the presence of water.

  • Sequence: Fmoc-ε-azido-Nle-OH
  • CAS-No.: 159610-89-6
  • Mol.weight: 394.43
  • SumFormula: C₂₁H₂₂N₄O₄
  • Bulk item no.: B-4135
  • Synonym(s): Fmoc-Lys(N₂)-OH
  • EINECS no.: -
  • MDL no.: MFCD13182319

For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Formula: C₂₁H₂₂N₄O₄
MW: 394,431 g/mol
CAS Number: 159610-89-6
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty