BACEB-3570

4037463.0001 4037463.0005
B-3570.0005BAEA 990.59 USD
B-3570.0005BA B-3570.0001BA
BACEB-3570
Sequence: 1-(Fmoc-amino)-cyclopentanecarboxylic acid
Synonym(s): Fmoc-Ac₅c-OH

The acid fluoride of the sterically hindered Ac₅c could be coupled to N-methylated amino acids in HFIP at 55°C.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR