Fmoc-D-biocytin

4035775.0250
B-3550.0250BAEA 461.98 USD
B-3550.0250BA
Fmoc-D-biocytin
Fmoc-D-biocytin

Sequence: Fmoc-D-Lys(biotinyl)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Bachem also offers resins preloaded with Nε-labeled Fmoc-lysine: Fmoc-Lys(biotinyl)-Wang resin (D-2705) and Fmoc-Lys(Dnp)-SASRIN resin (D-2450).

Formula: C₃₁H₃₈N₄O₆S
Storage Temperature: Freezer
MDL Number: MFCD00270742
CAS Number: 110990-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR