Fmoc-allo-Thr-OH

Synonyms: (2S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid

4027556.0001 4027556.0005
B-3100.0005BAEA 2015.76 USD
B-3100.0005BA B-3100.0001BA
Fmoc-allo-Thr-OH
Fmoc-allo-Thr-OH

Sequence: Fmoc-allo-Thr-OH


Bachem also offers phosphothreonine derivatives.
aThr derivatives.

Formula: C₁₉H₁₉NO₅
MW: 341.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318739
CAS Number: 146306-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR