Fmoc-D-Arg-OH hydrochloride

4027303.0001
B-3040.0001BAEA 316.92 USD
B-3040.0001BA
Fmoc-D-Arg-OH hydrochloride
Fmoc-D-Arg-OH hydrochloride

Sequence: Fmoc-D-Arg-OH · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₁H₂₄N₄O₄·HCl
MW: 432.90 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318732
CAS Number: 130752-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR