Fmoc-Glu-ODmab

Supplier: BACHEM AMERICAS INC

4027142.0001 4027142.0005
B-3005.0001BAEA 396.73 USD
B-3005.0001BA B-3005.0005BA
Fmoc-Glu-ODmab
Fmoc-Glu-ODmab

Sequence: Fmoc-Glu-ODmab


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


The ODmab group is an excellent protecting group for carboxylic acids. It is orthogonal to both acid (TFA) and 20% piperidine-labile protecting groups used in standard and automated peptide chemistry. Dmab esters are quantitatively cleaved with 2% hydrazine in DMF within minutes.

Formula: C₄₀H₄₄N₂O₈
Storage Temperature: Freezer
MDL Number: MFCD00797874
CAS Number: 172611-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR