Fmoc-Arg(NO2)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Arg(NO₂)-OHNα-Fmoc-Nomega-nitro-L-arginine

4028841.0025 4028841.0005
B-2785.0025BAEA 572.92 USD InStock
B-2785.0025BA B-2785.0005BA
Fmoc-Arg(NO2)-OH
Fmoc-Arg(NO2)-OH

Sequence: Fmoc-Arg(NO₂)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₁H₂₃N₅O₆
MW: 441,444 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065617
CAS Number: 58111-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR