Fmoc-Gln(1-adamantyl)-OH

4025248.0001
B-2435.0001BAEA 316.92 USD
B-2435.0001BA
Fmoc-Gln(1-adamantyl)-OH
Fmoc-Gln(1-adamantyl)-OH

Sequence: Fmoc-Gln(1-adamantyl)-OH


For isoglutamine derivatives please see the family "Glutamic Acid".

Formula: C₃₀H₃₄N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00237020
CAS Number: 159926-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR