Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Fmoc-Gly-Pro-Hyp-OH

Supplier: BACHEM AMERICAS INC

B-2080.0005 B-2080.0001
B-2080.0005BAEA 1194.79 USD InStock
B-2080.0005BA B-2080.0001BA
Fmoc-Gly-Pro-Hyp-OH
Proteins and Peptides
Sequence: Fmoc-Gly-Pro-Hyp-OH
CAS-No.: 185213-75-6
Mol.weight: 507.54
SumFormula: C₂₇H₂₉N₃O₇
Bulk item no.: B-2080
Synonym(s):
EINECS no.: -
MDL no.: MFCD00237658

For Fmoc-dipeptides Fmoc-Xaa-Gly-OH and Fmoc-Xaa-Pro-OH, which can be used as building blocks in Fmoc-SPPS, please see also the subfamily 'Fmoc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. Pseudoproline dipeptides and isoacyl dipeptides can be found in this section as well. In case of a strong tendency towards diketopiperazine formation during Fmoc-SPPS, Fmoc-dipeptides have been coupled even at the risk of a some epimerization.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty