Fmoc-d-asp(otbu)-opfp

Supplier: BACHEM AMERICAS INC

4016142.0001 4016142.0005
B-1845.0001BAEA 267.69 USD InStock
B-1845.0001BA B-1845.0005BA
Fmoc-d-asp(otbu)-opfp
Fmoc-d-asp(otbu)-opfp

Sequence: Fmoc-D-Asp(OtBu)-OPfp


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₉H₂₄F₅NO₆
MW: 577,504 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00153374
CAS Number: 200335-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR