(S)-1-Boc-piperidine-3-carboxylic acid

Synonyms: (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid1-Boc-L-nipecotic acid

4065479.0005
A-4760.0005BAEA 551.16 USD
A-4760.0005BA
(S)-1-Boc-piperidine-3-carboxylic acid
(S)-1-Boc-piperidine-3-carboxylic acid

Sequence: Boc-L-nipecotic acid


Nip and Inp derivatives.


Synonym(s): Boc-Nip-OH#Boc-(S)-piperidine-3-carboxylic acid

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02179172
CAS Number: 88495-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR