BACEA-4680
4037579.0100 4037579.0025
A-4680.0100BAEA 742.94 USD
A-4680.0100BA A-4680.0025BA
BACEA-4680
Sequence: Boc-Gly-OMe
Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
This Boc-protected glycine ester has been used in an N-heterocyclic carbene-catalyzed amidation as well as in the efficient synthesis of substituted oxopiperazines.