Boc-Dap(Cbz)-OH

Synonyms: Boc-Dap(Z)-OH(S)-2-[(tert-Butoxycarbonyl)amino]-3-(benzyloxycarbonylamino)propionic acid

4029288.0001
A-4675.0001BAEA 261.79 USD
A-4675.0001BA
Boc-Dap(Cbz)-OH
Boc-Dap(Cbz)-OH

Sequence: Boc-Dap(Z)-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.

Formula: C₁₆H₂₂N₂O₆
MW: 338.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275856
CAS Number: 65710-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR