BACEA-4645

4042361.0005 4042361.0001
A-4645.0001BAEA 309.56 USD
A-4645.0001BA A-4645.0005BA
BACEA-4645
Sequence: Boc-tBu-D-Gly-OH

A sterically demanding, lipophilic amino acid with a quaternary side-chain Cβ-atom. The bulky t-butyl group of Tle destabilizes α-helices.

Synonym(s): Boc-D-Tle-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR