BACEA-4615

4035503.0250 4035503.1000
A-4615.0250BAEA 793.47 USD
A-4615.0250BA A-4615.1000BA
BACEA-4615
Sequence: Boc-(Dmmb(Trt))Gly-OH

Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Synonym(s): Boc-N-(4,5-dimethoxy-2-tritylsulfanyl-Bzl)-Gly-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR